Learn More
Corning™ 384-Well Black, Clear Bottom Ultra-Low Attachment Microplate
Shop All Corning Products
Click to view available options
Color:
Black
Unit Size:
Pack of 100
Description
- Covalently bound hydrogel layer that effectively inhibits cellular attachment
- Surface minimizes protein absorption, enzyme activation and cellular activation
- Surface is noncytotoxic, biologically inert and nondegradable
- Sterilized by gamma irradiation and certified nonpyrogenic
- Black wall microplates have low background fluorescence, minimal light scatter and reduced crosstalk
- Opaque walls to prevent well-to-well crosstalk
- Optically clear flat well bottom permits direct microscopic viewing
- Bulk packed 20 per bag
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Culture Area | 0.16 cm2 |
| Array | 16 x 24 |
| Binding Type | Ultra-Low Attachment (covalently bound Hydrogel layer) |
| Color | Black |
| For Use With (Equipment) | Both top and bottom reading instruments |
| Material | Polystyrene |
| No. of Wells | 384 |
| Volume (Metric) Working | 20 to 80 μL |
| Show More |