Learn More
Corning™ 33mm Non-Vented Polyethylene Cap
BIOPHARMAGREENER_CHOICE
Product Code. 11333725 Shop All Corning Products
Change view
Click to view available options
Thread Size (Metric):
00 mm
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) unitSize
11333725 00 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11333725 Supplier Corning™ Supplier No. 10035
Only null left
Add to basket
Add to basket
Replacement cap for all High Yield Performance (HYPER) Flasks
This replacement cap can be used on all High Yield Performance HYPER Flasks (Cat. No. 10020, 10024, 10030, 10034)
  • 33mm threaded cap
  • Solid cap is used in closed systems or applications where venting is not necessary
  • Sterilized by gamma irradiation
Order Info
Bottle sold separately
TRUSTED_SUSTAINABILITY
Specifications
Color Orange
Material Polyethylene
No. per Case 4/Cs.
No. per Pack 1/Pk.
Sterility Sterile
Thread Size (Metric) 00 mm
Neck Finish 33
For Use With (Equipment) High yield performance hyper flask