Learn More
Fisherbrand™ Cork Borer
Product Code. p-8012536 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Outer:
6.25mm
7.5mm
8.75mm
10mm
11.25mm
12.5mm
13.75mm
17.50mm
26.25mm
Unit Size:
Each
Product Code. 12900896
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Manufactured from brass and nickel-plated.
  • Comes complete with push rod for clearing borers
  • All sizes are 100mm in length
  • Tube wall thickness of 0.45mm
Specifications
Product Type Cork Borer
Diameter (Metric) Outer 11.25mm
Material Nickel-plated brass