Learn More
Proteintech CoraLite™488-Annexin V and PI Apoptosis Kit
CoraLite™488-Annexin V and PI Apoptosis Detection Kit provides a quick and easy method to detect apoptosis by marking early apoptotic cells (green) and late apoptotic cells or necrotic cells (red).
Brand: Proteintech PF00005-100T
Additional Details : Weight : 0.01900kg
Description
- Results can be read out using flow cytometry or other fluorescence testing equipment.
- Early apoptic cells are identified by annexin V (labeled with CoraLite(R) 488 dye) binding to phosphatidylserine (PS) flipped to the extracellular membrane surface, an early event in apoptosis.
- Compared with FITC, CoraLite(R) 488 dye has higher fluorescence brightness, and is not affected by pH in the environment, and has good light stability.