Learn More
Tocris Bioscience™ CoraFluor™ 1, thiol reactive
missing translation for 'orderingAttributeHoverText'
Quantity:
20 μg
missing translation for 'unitSize'
20µg
Beschreibung
CoraFluor™ 1, thiol reactive is a terbium-based time-resolved Förster resonance energy transfer (TR-FRET) donor for TR-FRET assay development, such as target engagement and ternary complex assays. This product is designed with maleimide functionality to conjugate to cysteines, making it suitable for labeling biomolecules, including antibodies, nanobodies, proteins and peptides. It is exceptionally bright, providing unmatched assay sensitivity and stability in biological media over other commercially available TR-FRET donors. CoraFluor™ 1, thiol reactive is compatible with common acceptor dyes such as FAM, TMR and Cyanine 5, SE (Cat. No. 5436), enabling seamless assay integration in high throughput screening applications. CoraFluor™1, amine reactive and CoraFluor™2, amine reactive also available.
Spezifikation
Spezifikation
| Quantity | 20 μg |
| Target | Fluorescent Probes |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur