missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Sulphate solution EU Pharmacopoeia 0.25M, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Unit Size:
1000mL
Specifications
Specifications
| Quantity | 1000 mL |
| Grade | Eu. Ph. |
| Concentration or Composition (by Analyte or Components) | 0.25M |
| Packaging | Bottle |
| Chemical Name or Material | Copper Sulphate Solution - CuSO4. |
Safety and Handling
Product Identifier
- Copper sulphate 0,25M. Volumetric Solution (RP 280)
- LONG-TERM AQUATIC HAZARD Chronic 2
- H411-Toxic to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction