Learn More
Copper, Naval Brass, Uninhibited, CDA 464, C46400M, ARMI∣MBH
Description
Copper, Naval Brass, Uninhibited, CDA 464, C46400M, Ø40x20mm
Specifications
Specifications
| Chemical Name or Material | Manganese/Antimony/Silver/Arsenic/Phosphorus/Nickel/Iron/Lead/Tin/Zinc/Copper/Aluminium/Silicon |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Manganese 0.001 %/Antimony 0.0043 %/Silver 0.0043 %/Arsenic 0.0052 %/Phosphorus 0.0136 %/Nickel 0.015 %/Iron 0.0488 %/Lead 0.458 %/Tin 1.13 %/Zinc 37.07 %/Copper 61.24 %/Aluminium (0.0006) %/Silicon (0.0033) % |