Learn More
Hanna Instruments™ Copper LR Reagent Water Testing Kit
Click to view available options
Quantity:
100/Pk.
Unit Size:
Pack of 100
Specifications
Specifications
| No. of Tests | 100 |
| Quantity | 100/Pk. |
| Range | 0 to 1.5mg/L |
| Test Method | Biconchonate Acid |
| Type | Copper LR Reagent |
Safety and Handling
Product Identifier
- X100 TEST COPPER LR
- Danger
- Respiratory sensitiser Category 1A
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- MIXTURE LIST-Contains : bicinchonnic acid disodium salt