missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Leaded Yellow Brass, CDA 857, C85700, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Copper, Leaded Yellow Brass, CDA 857, C85700
Specifications
Specifications
| Chemical Name or Material | Aluminium, Antimony, Arsenic, Bismuth, Carbon, Chromium, Cobalt, Copper, Iron, Lead, Magnesium, Manganese, Nickel, Phosphorus, Silicon, Silver, Tin, Zinc |
| Diameter (Metric) Outside | 38 mm |
| Form | Disc |
| Thickness (Metric) | 3 mm |
| Concentration or Composition (by Analyte or Components) | Bismuth: 0.003%, Carbon: 0.003%, Silicon: 0.004%, Manganese: 0.006%, Arsenic: 0.007%, Cobalt: 0.007%, Phosphorus: 0.008%, Antimony: 0.014%, Nickel: 0.095%, Aluminium: 0.2%, Iron: 0.29%, Tin: 0.78%, Lead: 1.58%, Zinc: 36.1%, Copper: 60.9%, Chromium: 0.002%, Magnesium: 0.01%, Silver: 0.01% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction