Learn More
Copper(II) Sulfate Solution, Chem Lab™
Click to view available options
Quantity:
1 L
Unit Size:
Each
Specifications
Specifications
| Color | Blue |
| pH | ±4 |
| Physical Form | Liquid |
| Quantity | 1 L |
| Concentration or Composition (by Analyte or Components) | 0.01mol/L |
| Density | 1.01g/mL |