missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) acrylate, typically 95%, Thermo Scientific Chemicals

Product Code. p-7051688
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11328928 5 g 5g
11318928 25 g 25g
11348928 100 g 100g
11338928 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11328928 Supplier Thermo Scientific Alfa Aesar Supplier No. 041779.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20074-76-4
Molecular Formula C6H6CuO4
Molecular Weight (g/mol) 205.656
MDL Number MFCD00078371
InChI Key XPLSDXJBKRIVFZ-UHFFFAOYSA-L
Synonym copper acrylate, copper ii acrylate, 2-propenoic acid,copper 2+ salt 9ci, acmc-20aldy, acrylate,copper ii, copper 2+ diacrylate, copper 2+ ion diacrylate, bis acrylic acid copper ii salt, copper ii acrylate, typically
PubChem CID 13682633
IUPAC Name copper;prop-2-enoate
SMILES C=CC(=O)[O-].C=CC(=O)[O-].[Cu+2]

Specifications

Odor Weak
Quantity 5 g
Formula Weight 205.66
Percent Purity 95% (typically)
Physical Form Powder
Chemical Name or Material Copper(II) acrylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.