missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) oxide, 99.9% (metals basis)

Product Code. 11369327
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 11369327

Brand: Thermo Scientific Alfa Aesar 040189.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1317-39-1 | Cu2O

It is used chiefly as a pigment in ceramics and in antifouling paints.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-39-1
Molecular Formula Cu2O
MDL Number MFCD00010974

Specifications

Boiling Point 1800°C (decomposition)
Melting Point 1235°C
Color Purple-Red
Physical Form Powder
Quantity 100 g
Assay Percent Range 99% (Metal basis)
Solubility Information Insoluble in water but soluble in acids.
Formula Weight 143.08
Odor Odorless
Refractive Index 2.705
Sensitivity Air and moisture sensitive
Appearance Purple-red powder
Density 6
Chemical Name or Material Copper(I) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.