missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, High Silicon Bronze A, CDA 655, C65500, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, High Silicon Bronze A, CDA 655, C65500, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Carbon/Phosphorus/Silver/Chromium/Oxygen/Sulfur/Antimony/Magnesium/Arsenic/Palladium/Aluminium/Nickel/Tin/Lead/Iron/Zinc/Manganese/Silicon/Copper/Bismuth/Cadmium/Cobalt/Molybdenum/Boron/Hydrogen/Titanium/Calcium/Nitrogen/Selenium/Vanadium |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Carbon -100 ppm/Phosphorus -20 ppm/Silver -15 ppm/Chromium -10 ppm/Oxygen -10 ppm/Sulfur -10 ppm/Antimony -8 ppm/Magnesium -5 ppm/Arsenic -4 ppm/Palladium -0.3 ppm/Aluminium 0.002 %/Nickel 0.0034 %/Tin 0.009 %/Lead 0.018 %/Iron 0.055 %/Zinc 0.149 %/Manganese 0.98 %/Silicon 3.15 %/Copper 95.5 %/Bismuth (<10) ppm/Cadmium (<10) ppm/Cobalt (<10) ppm/Molybdenum (<10) ppm/Boron (<100) ppm/Hydrogen (<2) ppm/Titanium (<20) ppm/Calcium (<33) ppm/Nitrogen (<5) ppm/Selenium (<5) ppm/Vanadium (<50) ppm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction