Learn More
Copper Dipropanoate, TRC
Product Code. 30366880
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30366880 1 g 1g
30351891 5 g 5g
30355384 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30366880 Supplier Toronto Research Chemicals Supplier No. TRCC6853151G
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3112-74-1
Molecular Formula C6H10CuO4
Molecular Weight (g/mol) 209.69
Synonym Copper Dipropionate, Copper(2+) Propionate, Copper(II) Propanoate, Copper(II) Propionate, Cupric Propanoate, Cupric Propionate
IUPAC Name copper;propanoate
SMILES [Cu+2].CCC(=O)[O-].CCC(=O)[O-]
Specifications
Quantity 1 g
InChI Formula InChI=1S/2C3H6O2.Cu/c2*1-2-3(4)5;/h2*2H2,1H3,(H,4,5);/q;;+2/p-2
Formula Weight 208.9875
Chemical Name or Material Copper Dipropanoate