Learn More
Palintest™ Copper (Coppercol) Test Kit
Product Code. 11903617
Click to view available options
Quantity:
50 Pack
Unit Size:
Pack of 50
Product Code. 11903617
Brand: Palintest™ PM186
Specifications
No. of Tests 50
Quantity 50 Pack
Range 0 to 5mg/L
Sufficient For 50 Tests
Type Copper (Coppercol) Test Kit
compliance-icons
Product Identifier
  • photometry test kit copper 0-5PPM
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : disodium hydrogen phosphate, adipic acid, sodium metabisulphite, boric acid