Learn More
Thermo Scientific™ Copper Beaker with Pourout Lip
Product Code. p-6685013
Change view
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
2000 mL
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
15423695 125 mL Each
15433695 250 mL Each
15443695 500 mL Each
15463695 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15423695 Supplier Thermo Scientific™ Supplier No. 039052.KT
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Available in a range of capacities
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
  • Pourout lip allows for easy pouring
TRUSTED_SUSTAINABILITY
Specifications
Description With Pourout Lip
Diameter (Metric) Outer Top: 56 mm; Bottom: 48 mm
Capacity (Metric) 125 mL
Material Copper
Height (Metric) 67 mm
Type Beaker