missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Aluminum Bronze, CDA 954, C95400, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Aluminum Bronze, CDA 954, C95400, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Antimony/Bismuth/Chromium/Carbon/Lead/Phosphorus/Cobalt/Silicon/Tin/Nickel/Zinc/Manganese/Iron/Aluminium/Copper/Hafnium/Hydrogen/Palladium/Tantalum/Tungsten/Yttrium/Zirconium/Beryllium/Calcium/Niobium/Selenium/Titanium/Vanadium/Nitrogen/Boron/Oxygen/Magnesium/Cadmium/Molybdenum/Arsenic/Silver |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Antimony 0.0009 %/Bismuth 0.0011 %/Chromium 0.003 %/Carbon 0.007 %/Lead 0.016 %/Phosphorus 0.016 %/Cobalt 0.017 %/Silicon 0.025 %/Tin 0.047 %/Nickel 0.134 %/Zinc 0.141 %/Manganese 0.29 %/Iron 4.23 %/Aluminium 10.36 %/Copper 84.7 %/Hafnium (<0.001) %/Hydrogen (<0.001) %/Palladium (<0.001) %/Tantalum (<0.001) %/Tungsten (<0.001) %/Yttrium (<0.001) %/Zirconium (<0.001) %/Beryllium (<0.005) %/Calcium (<0.005) %/Niobium (<0.005) %/Selenium (<0.005) %/Titanium (<0.005) %/Vanadium (<0.005) %/Nitrogen (0.0006) %/Boron (0.0007) %/Oxygen (0.001) %/Magnesium (0.0013) %/Cadmium (0.002) %/Molybdenum (0.002) %/Arsenic (0.003) %/Silver (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction