missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, High Leaded Tin Bronze, CDA 937, C93700, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, High Leaded Tin Bronze, CDA 937, C93700, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Aluminium/Manganese/Silicon/Arsenic/Iron/Bismuth/Silver/Phosphorus/Antimony/Zinc/Nickel/Lead/Tin/Copper/Beryllium/Chromium/Cobalt/Selenium/Carbon/Oxygen |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Aluminium 0.0013 %/Manganese 0.0016 %/Silicon 0.0019 %/Arsenic 0.005 %/Iron 0.008 %/Bismuth 0.011 %/Silver 0.05 %/Phosphorus 0.073 %/Antimony 0.078 %/Zinc 0.146 %/Nickel 0.17 %/Lead 9.42 %/Tin 9.65 %/Copper 80.35 %/Beryllium (<0.001) %/Chromium (0.0007) %/Cobalt (0.0007) %/Selenium (0.001) %/Carbon (0.002) %/Oxygen (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction