missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, High Leaded Tin Bronze, CDA 932, C93200, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, High Leaded Tin Bronze, CDA 932, C93200, Ø38x19mm
Specifications
Specifications
| Chemical Name or Material | Aluminium/Cadmium/Oxygen/Cobalt/Carbon/Arsenic/Selenium/Bismuth/Silver/Sulfur/Phosphorus/Iron/Antimony/Nickel/Zinc/Tin/Lead/Copper/Germanium/Indium/Tellurium/Hafnium/Hydrogen/Magnesium/Manganese/Niobium/Nitrogen/Tantalum/Tungsten/Zirconium/Chromium/Beryllium/Boron/Calcium/Molybdenum/Silicon/Titanium |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Aluminium 0.0007 %/Cadmium 0.0007 %/Oxygen 0.002 %/Cobalt 0.0024 %/Carbon 0.0025 %/Arsenic 0.0073 %/Selenium 0.009 %/Bismuth 0.0094 %/Silver 0.019 %/Sulfur 0.031 %/Phosphorus 0.04 %/Iron 0.07 %/Antimony 0.31 %/Nickel 0.454 %/Zinc 3.44 %/Tin 6.82 %/Lead 7.95 %/Copper 81.2 %/Germanium (<0.0001) %/Indium (<0.0001) %/Tellurium (<0.0001) %/Hafnium (<0.0010) %/Hydrogen (<0.0010) %/Magnesium (<0.0010) %/Manganese (<0.0010) %/Niobium (<0.0010) %/Nitrogen (<0.0010) %/Tantalum (<0.0010) %/Tungsten (<0.0010) %/Zirconium (<0.0010) %/Chromium (<0.0040) %/Beryllium (<0.0050) %/Boron (<0.0050) %/Calcium (<0.0050) %/Molybdenum (<0.0050) %/Silicon (<0.0050) %/Titanium (<0.0050) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction