missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper, Leaded Commercial Bronze, CDA 314, C31400, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Copper, Leaded Commercial Bronze, CDA 314, C31400, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Carbon/Oxygen/Nickel/Phosphorus/Antimony/Iron/Tin/Lead/Zinc/Copper/Nitrogen/Silicon/Silver/Arsenic |
| CAS | 7440-50-8 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Carbon 0.002 %/Oxygen 0.002 %/Nickel 0.004 %/Phosphorus 0.005 %/Antimony 0.006 %/Iron 0.007 %/Tin 0.029 %/Lead 1.99 %/Zinc 7.81 %/Copper 90.08 %/Nitrogen (0.0002) %/Silicon (0.002) %/Silver (0.002) %/Arsenic (0.003) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction