Learn More
Epredia™ Coplin Staining Jar
Hold 10 slides back-to-back with the Epredia™ Plastic Coplin Staining Jar.
Brand: Epredia™ 194
Additional Details : Weight : 0.05760kg
Description
Features:- Holds 10 slides back-to-back
- 1.6 in. (40mm) diameter circular tapered top
- High-density polyethylene construction with internal grooves
Specifications
Plastic Coplin Staining Jar | |
Plastic | |
Coplin Staining Jar | |
Domed screw-on cap |
Not intended to be microwavable