Learn More
Berthold Technologies Cooling/Heating Unit for LED Plant Growth Illumination
Product Code. 31908714 Shop All Berthold Technologies Products
Change view
Click to view available options
For Use With (Application):
Plant in vivo Imaging
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
31908714 Plant in vivo Imaging Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31908714 Supplier Berthold Technologies Supplier No. 56695
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
To cool the LED panels of the NightSHADE LB 985.
  • The LED panels of the NightSHADE in vivo Imaging System for plants allow to simulate daylight, but they produce a considerable amount of heat.
  • This device cools the LED panels and prevents overheating.
Specifications
Type Cooling/Heating Unit
For Use With (Application) Plant in vivo Imaging
For Use With (Equipment) LB 985 NightSHADE