Learn More
Corning™ Extension Collar for CoolBox™ XT System
BIOPHARMA
Product Code. 15592851
Click to view available options
Color:
Green
Orange
Pink
Purple
Unit Size:
Each
Lid closure not possible with addition of extension collar
Product Code. 15592851
Brand: Corning™ 432084
Lid closure not possible with addition of extension collar
For use with CoolBox™ XT System. Corning™ Extension Collar is required when using taller CoolRack™ tube modules and tall tubes in CoolBox system.
For use with CoolBox XT and CoolBox 2XT to extend the height when using tall tube modules. The collar is magnetized and easily adheres to the unit base.
TRUSTED_SUSTAINABILITY
Specifications
Color Green
Shape Square
Type Extension Collar
For Use With (Application) Extend the Height When Using Tall Tube Modules
For Use With (Equipment) CoolBox XT and CoolBox 2XT
Quantity 1/Pk.
Dimensions (L x W x H) 20.0 x 16.0 x 7.0 cm (7.8 x 6.2 x 2.7 in.) Interior: 14.0 x 9.4 x 7.0 cm (5.5 x 3.7 x 2.7 in.)
Length (English) 7.8 in.
Length (Metric) 20 cm
Width (English) 6.2 in.
Width (Metric) 16 cm
Height (English) 2.75 in.
Height (Metric) 7 cm