Learn More
Masterflex™ Convoluted FEP Tubing
Product Code. 11736148
Click to view available options
Diameter (English) Inner:
1/4 in.
Unit Size:
Each
Product Code. 11736148
Brand: Masterflex™ 0648604
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Convoluted spiral design promotes full drainage and increases tubing flexibility
  • Chemical-resistant fluoropolymer tubing
  • Available in 5-ft lengths with 1 in. straight sections at both ends for easy connections
  • Choose from FEP or PFA formulations
  • Use tubing with barbed fittings
Specifications
Diameter (English) Inner 1/4 in.
Diameter (Metric) Inner 6.4 mm
Material FEP
Diameter (English) Outer 9/32 in.
Diameter (Metric) Outer 7.1 mm
Overall Length (English) 5 ft.
Overall Length (Metric) 1.5 m
Min. Operating Temperature (English) -450°F
Min. Operating Temperature (Metric) -270°C
Color Transparent
Maximum Pressure 56 psi
Max. Operating Temperature (English) 400°F
Max. Operating Temperature (Metric) 205°C
Product Type Convoluted FEP Tubing