missing translation for 'onlineSavingsMsg'
Learn More

Convicine, TRC

Product Code. 17937714
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17937714 10 mg 10mg
17957904 1 mg 1mg
17938504 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17937714 Supplier Toronto Research Chemicals Supplier No. TRCC68509510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19286-37-4
Molecular Formula C10 H15 N3 O8
Molecular Weight (g/mol) 305.24
Synonym 2,4(1H,3H)-Pyrimidinedione, 6-amino-5-(β-D-glucopyranosyloxy)-, Convicine (7CI,8CI), 6-Amino-5-(β-D-glucopyranosyloxy)-2,4(1H,3H)-pyrimidinedione, 6-Amino-5-(β-D-glucopyranosyloxy)-2,4-1H,3H-pyrimidinedione
IUPAC Name 6-amino-5-[(2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-1H-pyrimidine-2,4-dione
SMILES NC1=C(O[C@@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)C(=O)NC(=O)N1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C10H15N3O8/c11-7-6(8(18)13-10(19)12-7)21-9-5(17)4(16)3(15)2(1-14)20-9/h2-5,9,14-17H,1H2,(H4,11,12,13,18,19)/t2-,3-,4+,5-,9+/m1/s1
Formula Weight 305.0859
Percent Purity >95%
Chemical Name or Material Convicine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.