Learn More
Decon™ Contrad 2000 Cleaning Agent
Click to view available options
Volume (Metric):
5 L
Unit Size:
Each
Description
- Biodegradable
- Bactericidal
- Non-flammable
- Totally rinsable
- Combines highest quality anionic and non-ionic surface active agents with stabilising agents, alkalis, non-phosphate detergent builders and sequestering agents in an aqueous base.
- Highly alkaline (pH 13+) concentrate.
- Does NOT contain phosphates, enzymes, EDTA/NTA or chlorine bleaches.
- Safely and effectively cleans and decontaminates glassware, ceramics, most plastics, rubbers, stainless steel and ferrous metals.
- Not suitable for use on non-ferrous metals (notably aluminum and zinc), or on polycarbonate.
- Not suitable for domestic use
Specifications
Specifications
| Product Type | Cleaning Agent |
| pH | >13 |
| Appearance | Milky white |
| Boiling Point | 100.5°C |
| Certifications/Compliance | BS EN 1040 1997 for bacterial activity |
| Composition | A mixture of anionic and non-ionic surface active agents, stabilizing agents, alkalies, non-phosphat |
| Content And Storage | Store the concentrate at ambient temperature, out of direct sunlight, sealed in the container in which it is supplied, and away from strong acids and Aluminum. |
| Density | 1.083 at 20°C |
| Vapor Pressure | 17.1mmHg at 20°C |
| Volume (Metric) | 5 L |
Safety and Handling
Product Identifier
- contrad 2000
- Warning
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- H290-May be corrosive to metals.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : potassium hydroxide