Learn More
Contec™ VertiKlean™ Sox Mop Head Covers
BIOPHARMA
Product Code. 11976935 Shop All Contec Products
Click to view available options
Size:
Large
Medium
Regular
For Use With (Equipment):
Large VertiKlean Mop Head
Medium VertiKlean Mop Head
Regular VertiKlean Mop Head
Unit Size:
Pack of 120
Pack of 32
Product Code. 11976935
Brand: Contec™ VKSOX
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Increases durability of VertiKlean mop when used on abrasive surfaces. Contec™ VertiKlean™ Sox Mop Head Covers are available in regular, medium and large to fit all VertiKlean mop heads.
  • Clean-processed polyester
  • Extends the life of VertiKlean mop heads
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mop Cover
Sterility Non-sterile
Autoclavable Not Autoclavable
Size Regular
Disposable Single-use
For Use With (Equipment) Regular VertiKlean Mop Head
For Use With (Application) Removable cover increases durability of VertiKlean mop when used on abrasive surfaces
Material Polyester