Learn More
Contec™ VertiKlean™ Sox Mop Head Covers
BIOPHARMAGREENER_CHOICE
Product Code. 11966935 Shop All Contec Products
Click to view available options
Size:
Large
Medium
Regular
For Use With (Equipment):
Large VertiKlean Mop Head
Medium VertiKlean Mop Head
Regular VertiKlean Mop Head
Unit Size:
Pack of 120
Pack of 32
Product Code. 11966935
Brand: Contec™ MVKQTSOX
Increases durability of VertiKlean mop when used on abrasive surfaces. Contec™ VertiKlean™ Sox Mop Head Covers are available in regular, medium and large to fit all VertiKlean mop heads.
  • Clean-processed polyester
  • Extends the life of VertiKlean mop heads
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mop Cover
Color Code White
Sterility Non-sterile
Autoclavable Not Autoclavable
Size Medium
Disposable Single-use
For Use With (Equipment) Medium VertiKlean Mop Head
For Use With (Application) Removable cover increases durability of VertiKlean mop when used on abrasive surfaces
Color-coded No
Material Polyester