Learn More
Greiner Bio-One Contact Dishes
Product Code. p-7075395
Click to view available options
Vent:
No
Unit Size:
Pack of 600
Product Code. 10005090
Brand: Greiner Bio-One 629161
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Sterilised by irradiation. Greiner Bio-One Contact Dishes used in hygiene monitoring for detecting microorganisms and testing effectiveness of cleaning and disinfection on flat surfaces.
  • Available in different sizes
  • With or without vents
  • Material: Crystal clear and heat resistant polystyrene
  • Contact dish with graduation to enable quick and easy analyses
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Material Polystyrene
Shape Round
Sterility Sterile
Type Cell Culture Dish
Vent No
Height (Metric) 15 mm
Diameter (Metric) 65 mm