Learn More
Contec™ CONSTIX SP-6N Sealed Polyester Swabs
BIOPHARMA
Product Code. 15835204 Shop All Contec Products
Click to view available options
Length (Metric):
162.6 mm
Unit Size:
Pack of 500
Product Code. 15835204
Brand: Contec™ SP6N
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Contec™ CONSTIX SP-6N Sealed Polyester Swabs consists of a polyester nonwoven fabric that is thermally bonded to a flex-tip polypropylene handle.
  • Flexible tip provides a softer application on surfaces
  • Nonwoven polyester head provides enhanced sorbency for non-aqueous solutions
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Handle 3.0 mm
Diameter (Metric) Tip 2.5 mm
Length (Metric) Tip 16.3 mm
Length (Metric) 162.6 mm
Sterility Non-sterile
Width (Metric) Tip 6.9 mm
Handle Material Polypropylene