Learn More
Brady™ Consecutive Numbers & Letters, For Indoor Use
Product Code. 15523340
Change view
Click to view available options
Dimensions (L x W):
19 x 14 mm
38 x 22 mm
Unit Size:
Pack of 25
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15523340 19 x 14 mm Pack of 25
15573340 38 x 22 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15523340 Supplier Brady™ Supplier No. 034210
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Labels that can be permanent or repositionable.
  • Labels can be permanent or repositionable; once applied, numbers & letters are there to stay - unless you want to change them
  • Made of B-499 nylon cloth with pressure sensitive adhesive that sticks to any clean, dry surface
Specifications
Material Nylon Cloth (B-499)
Adhesive Type Permanent Acrylic
Thickness 0.166 mm
Dimensions (L x W) 19 x 14 mm
Color Black on Yellow