Learn More
SCAT Y-Connector
Product Code. p-7225784 Shop All SCAT Europe Products
Change view
Click to view available options
Diameter (Metric):
11 mm
3 mm
4 mm
5 mm
6 to 7 mm
9 mm
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) unitSize
10252760 3 mm Each
10374410 4 mm Each
10304180 5 mm Each
10346194 6 to 7 mm Each
10801361 9 mm Each
10651791 11 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10252760 Supplier SCAT Supplier No. 107801
Only null left
Add to basket
Add to basket
Safe connection of tubes of different sizes: directly and without technical aids. For safe collection of liquid HPLC waste, and many other applications in the laboratory!
SCAT™ connectors are available in the following versions:
  • Conical connectors
  • Y-connectors
  • Tube connectors
  • Capillary connectors
Specifications
Diameter (Metric) 3 mm
Autoclavable Autoclavable
Material Polypropylene
For Use With Tubing
For Use With (Application) HPLC/UHPLC Systems
Color Transparent
Product Type Y Connector