Learn More
SCAT™ PTFE Capillary
Product Code. 10314660 Shop All SCAT Europe Products
Click to view available options
Diameter (Metric):
1.6 mm
2.3 mm
3.2 mm
Unit Size:
3m
Product Code. 10314660
Brand: SCAT™ 461053
Safe connection of tubes of different sizes: directly and without technical aids. For safe collection of liquid HPLC waste, and many other applications in the laboratory!
SCAT™ connectors are available in the following versions:
  • Conical connectors
  • Y-connectors
  • Tube connectors
  • Capillary connectors
Specifications
Diameter (Metric) 1.6 mm
Autoclavable Autoclavable
Material PTFE
For Use With Standard Capillaries
For Use With (Application) HPLC/UHPLC Systems
Color Transparent
Product Type Capillary Connector