Learn More
Binder Connection Kit VDL VP5 Stainless Steel
Shop All BINDER Products
Click to view available options
For Use With:
VP5 Chemical Membrane Pump
Unit Size:
Each
Description
- Consisting of stainless steel hose 1.9 m long, with small flange 1/2 in. in diameter (DN 16) on both sides, two clamping rings and two universal centering rings.
Specifications
Specifications
| For Use With | VP5 Chemical Membrane Pump |
| IP Rating | IP52 |
| Tubing Size | DN 16 |
| Type | Pump Connection Kit |