Learn More
Binder Connection Kit VDL VP5 PTFE
Shop All BINDER Products
Click to view available options
For Use With:
Vacuum Pump VP5 EX
Unit Size:
Each
Description
- Consisting of antistatic PTFE hose 1.9 m long, with small flange 1/2 in. in diameter (DN 16) on both sides, two clamping rings and two universal centering rings.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| For Use With | Vacuum Pump VP5 EX |
| IP Rating | IP52 |
| Type | Pump Connection Kit |