Learn More
Binder Connection Kit VDL VP5 PTFE
Product Code. 30503387 Shop All BINDER Products
Click to view available options
For Use With:
Vacuum Pump VP5 EX
Unit Size:
Each
Product Code. 30503387
Brand: Binder 80122614
For vacuum pump VP5 EX.
  • Consisting of antistatic PTFE hose 1.9 m long, with small flange 1/2 in. in diameter (DN 16) on both sides, two clamping rings and two universal centering rings.
Specifications
Autoclavable Not Autoclavable
For Use With Vacuum Pump VP5 EX
IP Rating IP52
Type Pump Connection Kit