Learn More
WTW™ Connection Cable
104.00€ - 995.00€
Specifications
For Use With (Equipment) | MPP IDS Respectively IDS Sensors |
---|---|
Type | Connection Cable |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
15174155
|
WTW™
903850 |
1.5 m |
104.00€
1 unit |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
13476768
|
WTW™
903859 |
100 m |
995.00€
Each |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Specifications
MPP IDS Respectively IDS Sensors | |
Connection Cable |