Learn More
BRAND™ Conical Joint Stopper
Product Code. p-7164775 Shop All Brand Products
Click to view available options
Standard Taper Joint:
NS 10/19
NS 34/35
Unit Size:
Each
Product Code. 10653012
Brand: BRAND™ 144610
Conical Joint Stopper features a hollow design and hexagonal grip.
  • Borosilicate Glass 3.3
  • DIN 12252
  • Hollow with hexagonal grip
TRUSTED_SUSTAINABILITY
Specifications
Description Hollow with hexagonal grip
Standard Taper Joint NS 10/19
Material Borosilicate Glass
Shape Conical