Learn More
KARTELL Conical 1 Hole Stopper
Product Code. p-200004218
Click to view available options
Diameter (Metric) Top:
12 mm
13 mm
14 mm
18 mm
Unit Size:
Pack of 25
Product Code. 10378081
Brand: KARTELL 0382000
Only null left
Add to basket
Add to basket
The Conical 1 Hole Stopper is ideal to create a seal while attaching a hose to a flask or bottle.
  • General purpose laboratory red rubber stoppers.
  • Material: Red rubber
  • Autoclavable
Specifications
Diameter (Metric) Hole 3 mm
Quantity 25 pcs.
Autoclavable Autoclavable
Color Red
Diameter (Metric) Bottom 9 mm
Diameter (Metric) Outer 12 mm
Diameter (Metric) Top 12 mm
Material Rubber
Shape Conical
Sterility Non-sterile
Stopper Number 000