Learn More
Brady™ Cone™ Weather-resistant Blank Cones
Product Code. 11721935
Click to view available options
Color:
Orange
Yellow
Height (Metric):
635 mm
889 mm
Unit Size:
Each
Product Code. 11721935
Brand: Brady™ 077205
Only null left
Add to basket
Add to basket
Made of high impact, weather-resistant materials, the Cone™ Warning System is an ideal solution for your indoor or outdoor hazardous warning needs. Cone™ Weather-resistant Blank Cones are four-sided, so your message can't be missed.
  • Made of high impact, weather-resistant materials.
  • Ideal for your indoor or outdoor hazardous warning needs.
  • Cones are four-sided, so your message can't be missed.
Specifications
Alternate Part Number CONE-4SIDE-35H-YELLOW-BLANK
Displayed Text Blank
Height (English) 35 in.
Height (Metric) 889 mm
Length (English) 10.5 in.
Length (Metric) 266.7 mm
Packaging Quantity 1 Ea.
Color Yellow
Material Plastic
Type Blank Cone