Learn More
Hanna Instruments™ Conductivity Meter
Product Code. p-8005748
Click to view available options
Weight (Metric):
95 g
215 g
Unit Size:
Each
Product Code. 10182221
Brand: Hanna Instruments™ HI983304
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Temperature Compensation Automatic, 5 to 50°C (41 to 122°F) with β=2.4%/°C
Dimensions (L x W x H) 86 x 110 x 43 mm
Weight (Metric) 215 g
Warranty 2 Years
Probe Type HI 7631/2 with 2 m (6.6') cable and 1⁄2 in. thread(included).
Resolution 0.01 μS/cm
Power Supply 12 Vdc power adapter (included)
Type Conductivity Meter for Demineralized Water
Accuracy ±2% Full-scale