Learn More
BUCHI Condensers for BUCHI Rotavapors
Product Code. 12654709 Shop All Buchi Products
Change view
Click to view available options
Type:
Condenser S
Vertical Condenser V
Material (Coating):
P+G
Uncoated
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Material (Coating) unitSize
12654709 Vertical Condenser V P+G Each
12758380 Condenser S P+G Each
15600655 Vertical Condenser V Uncoated Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12654709 Supplier BUCHI Supplier No. 11055922
Only null left
Add to basket
Add to basket
Original BUCHI glassware guarantees the highest levels of efficiency and safety thanks to high precision in manufacturing. For more than 80 years, BUCHI has been manufacturing innovative and durable glass parts to ease daily laboratory work.
  • High throughput due to advanced design of condensers
  • Highly resistant glass provides the highest levels of safety (DURAN™ borosilicate glass 3.3)
  • Guaranteed leak-tightness and protection against hazardous fumes thanks to high precision joints
  • Maximum stability and safety with plastic and glass coating
TRUSTED_SUSTAINABILITY
Specifications
Material Borosilicate Glass (3.3)
Type Vertical Condenser V
For Use With (Equipment) Rotavapor R-210/R-215
Material (Coating) P+G