Learn More
Lenz Laborglasintrumente Condensers Acc. to Davies
Product Code. 10691702
Change view
Click to view available options
Standard Taper Joint No:
NS 24/29
NS 29/32
Length (Metric):
250 mm
400 mm
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Taper Joint No Length (Metric) Unit Size
10691702 NS 29/32 400 mm Each
10050381 NS 24/29 250 mm Each
10254101 NS 24/29 400 mm Each
10459172 NS 29/32 250 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10691702 Supplier Lenz Laborglasintrumente Supplier No. 05215404
  • ONLINE EXCLUSIVE PRICE
    185.00€
    128.84€ / Pack of 1
    Save 56.16€
    30% Off
Estimated Shipment: 12-01-2027
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Davies condensers for condensing vapours
  • With NS socket and NS cone
  • With double sized cooling area
  • Water connection: glass olives
  • Made of DURAN™ tubing
Specifications
Includes Glass olives
Length (Metric) 400 mm
Standard Taper Joint No NS 29/32
Material Borosilicate Glass (3.3)