Learn More
Contec™ CONSTIX™ Surface Validation Swabs
Product Code. p-3830794 Shop All Contec Products
Click to view available options
Length (Metric) Tip:
2 cm
2.31 cm
Unit Size:
Pack of 1000
Pack of 5
Product Code. 16685554
Brand: Contec™ SV2
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
CONSTIX™ Surface Validation Swabs designed for the evaluation of surface cleaning protocols in biopharmaceutical industries.
  • Package contains five polyester swabs specially processed to minimize background TOC contribution from the swab
  • Low background TOC content for minimal interference with test results
  • Large surface of the swab head and the long flexible handle provide exceptional surface coverage when swabbing
  • Strong, durable polyester swabs for optimum retrieval
  • Sterilizable by most common methods
  • Autoclavable
  • Swabs sealed in nylon pouch
TRUSTED_SUSTAINABILITY
Specifications
Diameter (English) Handle 0.12 in.
Length (English) 6 in.
For Use With (Application) Evaluation of surface cleaning protocols in bio-pharmaceutical industries
Product Type Swab
Diameter (Metric) Handle 3 mm
Length (Metric) 152.4 mm
Length (English) Handle 5.2 in.
Length (Metric) Handle 13.2 mm
Length (English) Tip 0.8 in.
Length (Metric) Tip 2 cm
Width (Metric) Tip 6.3 mm
Width (English) Tip 0.23 in.
Diameter (Metric) Tip 6.4 mm
Diameter (English) Tip 0.25 in.
Sterility Non-sterile
Adhesive Type Ultrasonically Sealed
Handle Material Polypropylene
Tip Style Round
Tip Material Knit Polyester