Learn More
Esculape Laboratoire™ Compress
Product Code. 11833922
Change view
Click to view available options
Dimensions (L x W):
300 x 300 mm
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
11833922 300 x 300 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11833922 Supplier Esculape Laboratoire™ Supplier No. 3170204
  • ONLINE EXCLUSIVE PRICE
    36.75€
    17.44€ / Pack of 100
    Save 19.31€
    52% Off
Estimated Shipment: 11-12-2026
Specifications
Product Type Gauze
Sterility Sterile
Length (Metric) 300 mm
Width (Metric) 300 mm
Dimensions (L x W) 300 x 300 mm