Description
Accommodate up to 19 slides with the Thermo Scientific™ Complete Staining Assembly 100, featuring a non-autoclavable glass dish.
Includes:
- 109 rack/handle
- 102 dish
- 101 cover
Staining Rack 109:
Made from stainless steel and holds up to 19 slides. Maximum slide size is 3 x 2 in. (75 x 50mm).
Staining Dish 102:
Made from non-autoclavable glass with inside dimensions of 3.5 x 2.75 x 2.55 in. (90 x 70 x 65mm). Holds up to 250mL of solution with a fully loaded staining rack.
Staining Dish Cover 101:
Made from stainless steel.
Specifications
Specifications
| Includes | Complete Assembly 100 includes 109 Rack/Handle, 102 Dish, and 101 Cover. |
| Product Type | Staining Dish Cover |