Learn More
JULABO™ Complete Carrier Trays with Test Tube Rack
Click to view available options
Holds:
240 Tubes, 100 x 16/17 mm dia.
360 Test Tubes, 12/13 mm dia.
360 Tubes, 30 mm x 11/12 mm dia.
Unit Size:
Each
Description
- Tray with 4 tube racks
- Metal construction
- Withstands temperatures up to 80 °C
- For use with JULABO™ shaking water baths SW22 / SW23
Specifications
Specifications
| For Use With (Equipment) | SW22, SW23 shaking water bath |
| Type | Carrier Tray |
| Holds | 240 Tubes, 100 x 16/17 mm dia. |
| Includes | 4 Racks |