Learn More
JULABO™ Complete Carrier Trays with Test Tube Rack
Product Code. 10140751
Click to view available options
Holds:
240 Tubes, 100 x 16/17 mm dia.
360 Test Tubes, 12/13 mm dia.
360 Tubes, 30 mm x 11/12 mm dia.
Unit Size:
Each
Product Code. 10140751
Brand: JULABO™ 8960440
Only null left
Add to basket
Add to basket
Includes 4 tube racks. Withstands temperatures up to 80°C. For use with JULABO™ shaking water baths SW22 / SW23.
  • Tray with 4 tube racks
  • Metal construction
  • Withstands temperatures up to 80 °C
  • For use with JULABO™ shaking water baths SW22 / SW23
Specifications
For Use With (Equipment) SW22, SW23 shaking water bath
Type Carrier Tray
Holds 240 Tubes, 100 x 16/17 mm dia.
Includes 4 Racks