Learn More
UVP Compact UV Lamp
Specifications
Specifications
| Voltage | 230 V |
| Wattage | 4 W |
| Wavelength | 365 nm |
| Length (Metric) | 198 mm |
| Material | Plastic |
| Model | UVL-23RS |
| Width (Metric) | 71 mm |
| Product Type | Financial UV Lamp |