Learn More
Nabertherm™ Compact Tube Furnace
Product Code. p-7187348 Shop All Nabertherm Products
Click to view available options
Dimensions (L x W x H) Exterior:
434 x 340 x 508 mm
670 x 340 x 508 mm
670 x 410 x 578 mm
920 x 460 x 628 mm
1170 x 460 x 628 mm
Controller Type:
B410
C450
P480
Temperature (Metric):
1200°C
1300°C
Unit Size:
Each
Product Code. 15456582
Brand: Nabertherm™ R151C2AN
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
These compact tube furnaces with integrated control systems can be used universally for many processes. Equipped with a standard working tube of C 530 ceramic and two fiber plugs, these tube furnaces have an unbeatable price/performance ratio.
  • Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
  • Solid state relays provide for low-noise operation
Specifications
Controller Type C450
Heated Zone 250 mm
Temperature (Metric) 1200°C
Dimensions (L x W x H) Exterior 434 x 340 x 508 mm
Length (Metric) 450 mm
Diameter (Metric) Process Tube 50 mm