Learn More
Hanna Instruments™ Compact Thermohygrometer
Product Code. 10535709
Click to view available options
Includes:
Dew Point, Calib. Data-Log Probe
with Calibration Data-Logging Probe
Unit Size:
Each
Product Code. 10535709
Brand: Hanna Instruments™ HI9564
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Type Compact Thermohygrometer
Resolution 0.1°C
Temperature Range (Metric) 0°C to 60°C
Temperature Accuracy ±3°C
Temperature Resolution 0.1°C
Weight (Metric) 340 g
Electrical Requirements 9V
Voltage 9 V
Includes with Calibration Data-Logging Probe